Skip to Content
Merck
All Photos(1)

Key Documents

880347P

Avanti

14:0-14:0(COOH) PC

Avanti Research - A Croda Brand 880347P, powder

Synonym(s):

1-myristoyl-2-(14-carboxymyristoyl)-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H70NO10P
CAS Number:
Molecular Weight:
707.92
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 100 mg (880347P-100mg)
pkg of 1 × 25 mg (880347P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880347P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCCCCCCCC(O)=O)=O)COC(CCCCCCCCCCCCC)=O)=O

Application

14:0-14:0(COOH) PC is suitable for use in the synthesis of phospholipid-nonaarginine conjugates (PL9R).

Biochem/physiol Actions

14:0-14:0(COOH) PC can be used to form well organized and stable phospholipid self-assembled monolayers (SAMs) by covalently attaching to silica substrates.

Packaging

5 mL Clear Glass Sealed Ampule (880347P-100mg)
5 mL Clear Glass Sealed Ampule (880347P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biomembrane mimetic surfaces by phospholipid self-assembled monolayers on silica substrates
Yang Z and Yu H
Langmuir, 15(5), 1731-1737 (1999)
Ji Hee Kang et al.
Biomacromolecules, 18(11), 3733-3741 (2017-09-28)
Despite potential applications of nucleic acid therapeutics, the lack of effective delivery systems hinders their clinical application. To overcome the barriers to nucleic acid delivery, we previously reported nanoparticles using phospholipid-polyethylenimine conjugates. However, toxicity of polyethylenimine remains as a problematic
Patrick Drücker et al.
Langmuir : the ACS journal of surfaces and colloids, 30(49), 14877-14886 (2014-11-22)
Solid-supported lipid bilayers (SLBs) mimicking a biological membrane are commonly used to investigate lipid-lipid or lipid-protein interactions. Simple binary or ternary lipid systems are well established, whereas more complex model membranes containing biologically important signaling lipids such as phosphatidylinositol-4,5-bisphosphate (PI(4,5)P2)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service