Skip to Content
Merck
All Photos(1)

Key Documents

700190M

Avanti

Cholesteryl(d7) linoleate

Avanti Research - A Croda Brand

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H69D7O2
CAS Number:
Molecular Weight:
656.13
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

methylene chloride solution

packaging

pkg of 1 × 1 mL (700190M-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

1 mg/mL (700190M-1MG)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](CC[C@]1([C@](CC=C2C[C@H](CC[C@]32C)OC(CCCCCCC/C=C\C/C=C\CCCCC)=O)4[H])[H])([C@]1(CC[C@@]43[H])C)[C@H](C)CCCC([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]

General description

Cholesteryl linoleate (LA-CE) is a major polyunsaturated fatty acid-containing cholesterol ester present in the core of low density lipoprotein (LDL).[1]

Biochem/physiol Actions

Cholesteryl linoleate (LA-CE) possesses antibacterial property.[2]

Packaging

2 mL Amber Glass Sealed Ampule

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Endogenous cholesterol ester hydroperoxides modulate cholesterol levels and inhibit cholesterol uptake in hepatocytes and macrophages
Guo S et al.
Redox Biology, 21, 101069-101069 (2019)
Lipids including cholesteryl linoleate and cholesteryl arachidonate contribute to the inherent antibacterial activity of human nasal fluid
Do Thai Q, et al.
Journal of Immunology, 181(6), 4177-4187 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service