Skip to Content
Merck
All Photos(1)

Key Documents

ATC311765799

Atlantic Research Chemicals Ltd

Ethyl Difluoro(6-methylpyridin-2-yl)acetate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
215.2
MDL number:

Iupac Name

ethyl difluoro(6-methyl-2-pyridinyl)acetate

Inchi Code

1S/C10H11F2NO2/c1-3-15-9(14)10(11,12)8-6-4-5-7(2)13-8/h4-6H,3H2,1-2H3

InChI key

PQERDYNTMXQQCR-UHFFFAOYSA-N

Purity

95

Country of Origin

GB

Product Link

https://atlantic-chemicals.com/product/?code=ES00278

MSDS

https://atlantic-chemicals.com/product/?code=ES00278


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library