Skip to Content
Merck
All Photos(1)

Key Documents

ATEH3031ECA7

AstaTech, Inc.

5-(BENZYLOXY)INDOLIN-2-ONE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
239.27
MDL number:

Iupac Name

5-(benzyloxy)-1H-indol-2-ol

Inchi Code

1S/C15H13NO2/c17-15-9-12-8-13(6-7-14(12)16-15)18-10-11-4-2-1-3-5-11/h1-9,16-17H,10H2

InChI key

DPPCWLOGRQMNRM-UHFFFAOYSA-N

Purity

95%

Physical Form

Liquid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=139276

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=AC2005


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library