APOH0D2000D9
Apollo Scientific Ltd
(4-(Difluoromethoxy)pyridin-2-yl)methanol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
(4-(difluoromethoxy)pyridin-2-yl)methanol
Inchi Code
1S/C7H7F2NO2/c8-7(9)12-6-1-2-10-5(3-6)4-11/h1-3,7,11H,4H2
InChI key
NZSFTRSYJIFQBH-UHFFFAOYSA-N
storage temp.
Ambient Temperature
Country of Origin
GB
Shipping Temperature
Ambient Temperature
Physical Form
Solid
Product Link
https://store.apolloscientific.co.uk/product/4-difluoromethoxypyridin-2-ylmethanol
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://store.apolloscientific.co.uk/product/4-difluoromethoxypyridin-2-ylmethanol
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.