Skip to Content
Merck
All Photos(1)

Documents

APO945298575

Apollo Scientific Ltd

Methyl 2-(3-methoxyphenyl)acetate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
180.2
MDL number:

Iupac Name

methyl (3-methoxyphenyl)acetate

Inchi Code

1S/C10H12O3/c1-12-9-5-3-4-8(6-9)7-10(11)13-2/h3-6H,7H2,1-2H3

InChI key

BSVIOYCZTJRBDB-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

96%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Liquid

Product Link

https://store.apolloscientific.co.uk/product/methyl-2-3-methoxyphenylacetate

Flash Point

86-88

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/methyl-2-3-methoxyphenylacetate


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service