APO456876691
Apollo Scientific Ltd
2-Fluoro-4-(trifluoromethyl)cinnamic acid
About This Item
Iupac Name
(2E)-3-[2-fluoro-4-(trifluoromethyl)phenyl]-2-propenoic acid
Inchi Code
1S/C10H6F4O2/c11-8-5-7(10(12,13)14)3-1-6(8)2-4-9(15)16/h1-5H,(H,15,16)/b4-2+
InChI key
HWBLGORXWYIISR-DUXPYHPUSA-N
storage temp.
Purity
Country of Origin
Shipping Temperature
Physical Form
Product Link
MSDS
https://store.apolloscientific.co.uk/product/2-fluoro-4-trifluoromethylcinnamic-acid
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.