Skip to Content
Merck
All Photos(1)

Key Documents

AMBH990F7551

Ambeed, Inc.

(6-(Trifluoromethyl)pyridin-2-yl)methanamine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
176.14
MDL number:

Iupac Name

[6-(trifluoromethyl)-2-pyridinyl]methanamine

Inchi Code

1S/C7H7F3N2/c8-7(9,10)6-3-1-2-5(4-11)12-6/h1-3H,4,11H2

InChI key

JDJHGZMTASXPEO-UHFFFAOYSA-N

storage temp.

Keep in dark place,Inert atmosphere,2-8C

Purity

98%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

Liquid or Semi-Solid or Solid or Lump

Product Link

https://www.ambeed.com/products/916304-19-3.html

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library