Skip to Content
Merck
All Photos(1)

Documents

Z756350

Nalgene® labware value pack start up kit

including Griffin beakers, graduated cylinders, wash bottles, jars, and racks

Synonym(s):

graduated cylinders, griffin beakers, griffin low form beakers, unwire racks

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121803
NACRES:
NB.43

manufacturer/tradename

Thermo Scientific 1350-0001

General description

All items in the labware value pack are chemically compatible with a wide range of common laboratory reagents; especially acids, bases and alcohols.
Pack Contents:
  • Straight-sided jars, PPCO, 500 mL (6)
  • Unitary wash bottles, LDPE, 500 mL (4)
  • Graduated cylinders, PPCO, 100 mL (2)
  • Graduated cylinders, PPCO, 500 mL (1)
  • Graduated cylinders, PPCO, 1 L (1)
  • Griffin low form beaker, PPCO, 50 mL (1)
  • Griffin low form beaker, PPCO, 100 mL (1)
  • Griffin low form beaker, PPCO, 250 mL (1)
  • Griffin low form beaker, PPCO, 600 mL (1)
  • Griffin low form beaker, PPCO, 1000 mL (1)
  • Unwire half-rack, blue for 30 mm tubes, RMT (1)

All components are autoclavable except for the Unitary wash bottles.

*PPCO= Polypropylene Copolymer
*LDPE= Low Density Polyethylene
*RMT= Resmer Manufacturing Technology

Compliance: Graduated Cylinders Meet CFR 21, Part 177.1520 and ASTM* E 1272 Class B accuracy requirements as well as ISO Standard 6706. Beakers meet ISO/DIN 7056 International Standards for Laborator Plasticware Accuracy.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Unwire is a trademark of Thermo Fisher Scientific or its subsidiaries

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service