Skip to Content
Merck
All Photos(1)

Documents

Z743725

ART® Low Retention Barrier Tip

ART 1000, size 1000 μL, racked, sterile

Synonym(s):

aerosol barrier tips, barrier filter tips, barrier tip, filter pipette tip, filter tip, pipet tip, sterile barrier pipette tip

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121603
NACRES:
NB.02

sterility

sterile

feature

ART 1000
racked

packaging

pkg of 4X800 ea

manufacturer/tradename

Molecular BioProducts 2279-05

technique(s)

PCR: suitable

size

1000 μL

Looking for similar products? Visit Product Comparison Guide

General description

  • ART® low retention barrier tips are designed to maximize sample recovery.
  • Tips hydrophobic inner surface prevents sample loss, facilitates sample accuracy and reproducibility.
  • Clear tips provide sample clarity.
  • Self-sealing technology prevents cross-contamination.
  • Certified RNase, DNase, ATP, endotoxin, and pyrogen free.

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service