Key Documents
Z743273
Julabo® adaptors
barbed fittings, for 12 mm i.d. tubing, pack of 2
Synonym(s):
barbed adaptor
Select a Size
2 EA
€27.20
€27.20
Please contact Customer Service for Availability
Select a Size
Change View
2 EA
€27.20
About This Item
UNSPSC Code:
41103706
NACRES:
NB.77
€27.20
Please contact Customer Service for Availability
packaging
pack of 2
manufacturer/tradename
Julabo 8970445
General description
For use with Julabo® circulators, 2 barbed fittings for tubing 12 mm ID. For use with M16x1 fittings. Requires a female M16x1 cap nut.
Legal Information
Julabo is a registered trademark of Julabo USA, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)