description
Designed to meet ISO 7027
feature
CE compliant
battery included
WEEE compliant
packaging
pkg of 1 ea
manufacturer/tradename
Extech Instruments TB400
measuring range
0-1000, resolution: 0.01 (Nephelometric Turbidity Units)
size
6.1 in. (155 mm) × 3.0 in. (76 mm) × 2.4 in. (62 mm)
weight
11.3 oz (320 g)
battery power
6 AAA
Features and Benefits
- Portable tester of water, food, beverages and other aqueous solutions.
- Meter comes complete with all the accessories to conveniently test the turbidity of municipal water, food and beverage water, and other aqueous solutions where fluid clarity is important. Portable design allows you to take measurement on-site instead of bringing solution back to the lab. Measures up to 1000(Nephelometric Turbidity Unit) with 0.01 NTU resolution.
- Microprocessor-based circuitry assures high accuracy and repeatable readings
- Requires only a 10mL sample size
- 2-point calibration
- Splash-proof front panel
- Battery operated for field and on-site testing
- Data Hold and Min/Max record/recall
- Complete with 0 NTU standard solution test bottle,100 NTU standard solution test bottle, cleaning solution (distilled water), (2) test bottles and covers, cleaning cloth, six AAA batteries, and hard carrying case
Choose from one of the most recent versions:
Certificates of Analysis (COA)