Key Documents
Z675881
Talboys dual block heaters
AC/DC input 230 V AC, UK plug
About This Item
UNSPSC Code:
41102404
NACRES:
NB.73
material
aluminum plate
manufacturer/tradename
Troemner 949513 UK
capacity
, holds 2 blocks
General description
This multi-purpose unit is ideal for incubation and activation of cultures, enzyme reactions, immunoassays, melting/boiling points and a wide variety of other laboratory procedures.
Economical, versatile and compact. It accepts 2 separate interchangeable modular heating blocks, which accommodate various sample enclosures such as microtubes, centrifuge tubes, vials, microplates, and PCR strips or tubes.
Each block has a thermometer well for measuring block temperature. Heats from ambient to 150 °C with two temperature adjustments, low range ambient to 100 °C/high range 75 °C to 150 °C. Takes only 50 mins to heat up to 100 °C.
The anodized aluminum modular heating blocks provide superior temperature stability and heat transfer.
  • Compact and economical
  • Holds 2 interchangeable heating blocks for maximum versatility
  • Heating blocks sold separately
Choose from one of the most recent versions:
Certificates of Analysis (COA)