Key Documents
Z547883
Sigma-Aldrich
SafetyBarb® hose connector
angled
Select a Size
1 EA
€16.80
€16.80
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€16.80
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
€16.80
Please contact Customer Service for Availability
General description
Consists of PBT cap, PP barbed connector 8.5 mm O.D., and silicone rubber seal. GL14 thread.
Legal Information
SafetyBarb is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)