Key Documents
Z507121
Supelco
Michel-Miller column plug
Front seal, PTFE, thread size 80 mm
Select a Size
1 EA
€311.00
€311.00
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
€311.00
About This Item
UNSPSC Code:
12000000
NACRES:
SB.52
€311.00
Please contact Customer Service for Availability
Request a Bulk Order
material
PTFE
manufacturer/tradename
Ace Glass 584660
technique(s)
LPLC: suitable
thread size
80 mm
General description
Solid plug for use as stopper with Ace-Threds. Seals M-M column ends, Plugs with #50 Ace-Thred fit Ace large columns.
Choose from one of the most recent versions:
Certificates of Analysis (COA)