Documents
Z281506
Sigma-Aldrich
Micro-90® concentrated cleaning solution
About This Item
UNSPSC Code:
47131800
General description
Cleaning methods include CIP, Manual and Immersion Washing, Mild-Agitation, Ultrasonic. Typical pH (neat) 9.0 to 9.9. Five-year shelf life. A mild yet powerful multi-purpose cleaner that is effective in both industrial and critical cleaning applications. Its unique formulation of chelants, ionic, and non-ionic ingredients produces a variety of cleaning actions that include lifting, dispersing, emulsifying, sequestering, suspending and decomposing. Excellent for cleaning soils include oil, grease, wax, tar, flux, particulates, and hard water stains. Outstanding for cleaning labware, ceramics, stainless steel, and defouling filter membranes.
Concentrate: dilute to 2% Micro in water.
May etch aluminum and zinc.
Legal Information
Micro-90 is a registered trademark of International Products
Visit the Document Library
Customers Also Viewed
Dimitrios A Koutsouras et al.
Materials science & engineering. C, Materials for biological applications, 81, 84-89 (2017-09-10)
Continuous and long-term monitoring of cellular and micro-organ activity is required for new insights into physiology and novel technologies such as Organs-on-Chip. Moreover, recent advances in stem cell technology and especially in the field of diabetes call for non-invasive approaches
Hugh L MacIntyre et al.
Journal of applied phycology, 30(2), 1073-1094 (2018-05-15)
Ballast water management systems (BWMS) must be tested to assess their compliance with standards for the discharge of organisms, for example in the ≥ 10- and < 50-μm size category, which is dominated by phytoplankton. Assessment of BWMS performance with the vital