Documents
Z278408
Sigma-Aldrich
Heat transfer block
Four holes for vials
About This Item
UNSPSC Code:
41102404
NACRES:
NB.43
feature
Holds 4 x flasks
manufacturer/tradename
Ace Glass 9594-12
General description
Four holes for all Ace microscale reaction vials and one hole for a thermometer. 27/8in. square × 3/4in. thick.
Aluminum blocks have excellent heat transfer rates and will not interfere with magnetic stirring. Replaces glass sand baths in microscale experiments. Use on top of stirring/hot plates.