Key Documents
Z267147
Sartorius balance analytical
Model BP300S, AC/DC input 220 V AC
About This Item
UNSPSC Code:
41111502
Pricing and availability is not currently available.
manufacturer/tradename
Model BP300S
General description
  • Large LCD readout
  • Push button internal calibration
  • Large, glass draft chamber
  • ISO/GLP compliance recording
  • RS232 serial interface
  • Overload protection
  • Level indicator
  • Lug for antitheft locking
  • CE mark of conformity
The industry standard for weighing tasks requiring fast, accurate results. Fully automatic calibration on all models ensures results are perfectly accurate with the press of a key. Convenient application programs offer parts counting, weighing in %, net total formulation, and mass unit conversions.
Choose from one of the most recent versions:
Certificates of Analysis (COA)