WHA10418312
Whatman® Nuclepore™ Track-Etched Membranes
polycarbonate filter, filter diam. 47 mm, pore size 3 μm, non-sterile, case of 100 ea
All Photos(1)
About This Item
Recommended Products
material
polycarbonate filter
sterility
non-sterile
packaging
case of 100 ea
manufacturer/tradename
Cytiva 10418312
filter diam.
47 mm
pore size
3 μm
Application
- Epifluorescence microscopy
- Environmental analysis
- Cell biologyEPA testing
- Fuel testing
- Bioassays
- Parasitology
- Air analysis
- Water microbiology
Legal Information
Nuclepore is a trademark of Cytiva
Whatman is a registered trademark of Cytiva