Skip to Content
Merck
All Photos(1)

Key Documents

W503304

Sigma-Aldrich

Furfuryl heptanoate

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18O3
CAS Number:
Molecular Weight:
210.27
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.131

biological source

synthetic

Quality Level

Assay

≥98%

refractive index

n20/D 1.459 (lit.)

density

1.001 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Organoleptic

green

SMILES string

CCCCCCC(=O)OCc1ccco1

InChI

1S/C12H18O3/c1-2-3-4-5-8-12(13)15-10-11-7-6-9-14-11/h6-7,9H,2-5,8,10H2,1H3

InChI key

VZRGLBPRLIGTPE-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lucie Froissart et al.
PloS one, 12(5), e0177581-e0177581 (2017-05-13)
Differences in learning and memory dynamics between populations are suspected to result from differences in ecological constraints such as resource distribution. The two reproductive modes (strains) of the parasitoid wasp Venturia canescens share the same geographical areas but live in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service