Skip to Content
Merck
All Photos(1)

Key Documents

W369101

Sigma-Aldrich

Hexyl benzoate

FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CO2(CH2)5CH3
CAS Number:
Molecular Weight:
206.28
FEMA Number:
3691
EC Number:
Council of Europe no.:
645
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.768
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA

refractive index

n20/D 1.493 (lit.)

bp

272 °C (lit.)

density

0.98 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

balsam; green; woody

SMILES string

CCCCCCOC(=O)c1ccccc1

InChI

1S/C13H18O2/c1-2-3-4-8-11-15-13(14)12-9-6-5-7-10-12/h5-7,9-10H,2-4,8,11H2,1H3

InChI key

UUGLJVMIFJNVFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suvi-Katriina Ruokonen et al.
Journal of chromatography. A, 1479, 194-203 (2016-12-14)
Liposomes were used as biomimetic models in capillary electrokinetic chromatography (EKC) for the determination of distribution constants (K
Eriko Harada et al.
PloS one, 7(1), e29710-e29710 (2012-01-13)
Odorant-binding proteins (OBPs) are extracellular proteins found in insect chemosensilla, where they participate in the sensing of odors, tastes, and pheromones. Although a large number of OBP genes have been identified in insect genomes, their molecular functions and biological roles
Filip Duša et al.
Electrophoresis, 37(2), 363-371 (2015-10-16)
The use of bare fused silica capillary in CE can sometimes be inconvenient due to undesirable effects including adsorption of sample or instability of the EOF. This can often be avoided by coating the inner surface of the capillary. In
Joanna Witos et al.
Journal of separation science, 43(12), 2495-2505 (2020-04-01)
Amphiphilic diblock copolymers consisting of a hydrophobic core containing a polymerized ionic liquid and an outer shell composed of poly(N-isoprolylacrylamide) were investigated by capillary electrophoresis and asymmetrical flow-field flow fractionation. The polymerized ionic liquid comprised poly(2-(1-butylimidazolium-3-yl)ethyl methacrylate tetrafluoroborate) with a
Nicholas R Larson et al.
Journal of medical entomology, 57(1), 187-191 (2019-09-10)
Common bed bug Cimex lectularius (L.) (Hemiptera: Cimicidae) infestations are on the rise and due to the development of pesticide resistance they are becoming more difficult to control, affordably. We evaluated a naturally occurring compound methyl benzoate (MB) and related

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service