W328804
2-Heptanol
≥97%
Synonym(s):
Methyl pentyl carbinol
About This Item
Recommended Products
biological source
synthetic
grade
Halal
Kosher
Assay
≥97%
refractive index
n20/D 1.420 (lit.)
bp
160-162 °C (lit.)
density
0.817 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
food allergen
no known allergens
Organoleptic
oily; earthy
SMILES string
CCCCCC(C)O
InChI
1S/C7H16O/c1-3-4-5-6-7(2)8/h7-8H,3-6H2,1-2H3
InChI key
CETWDUZRCINIHU-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Eye Irrit. 2 - Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
138.2 °F
Flash Point(C)
59 °C
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service