Skip to Content
Merck
All Photos(1)

Key Documents

W285900

Sigma-Aldrich

Phenethyl anthranilate

≥98%, FCC

Synonym(s):

Benzyl carbinyl anthranilate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(H2N)C6H4CO2CH2CH2C6H5
CAS Number:
Molecular Weight:
241.29
FEMA Number:
2859
EC Number:
Council of Europe no.:
258
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.723

biological source

synthetic

grade

Kosher

reg. compliance

FCC
FDA 21 CFR 172.515

Assay

≥98%

bp

226 °C (lit.)

mp

39-44 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

chocolate; honey; iris; fruity; rose; sweet

SMILES string

Nc1ccccc1C(=O)OCCc2ccccc2

InChI

1S/C15H15NO2/c16-14-9-5-4-8-13(14)15(17)18-11-10-12-6-2-1-3-7-12/h1-9H,10-11,16H2

InChI key

PXWNBAGCFUDYBE-UHFFFAOYSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service