Skip to Content
Merck
All Photos(1)

Key Documents

W263818

Sigma-Aldrich

Linalyl benzoate

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H22O2
CAS Number:
Molecular Weight:
258.36
FEMA Number:
2638
EC Number:
Council of Europe no.:
654
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.771

biological source

synthetic

Quality Level

grade

Halal
Kosher

Assay

≥95%

refractive index

n20/D 1.508 (lit.)

bp

213 °C (lit.)

density

0.98 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

floral

SMILES string

C\C(C)=C\CCC(C)(OC(=O)c1ccccc1)C=C

InChI

1S/C17H22O2/c1-5-17(4,13-9-10-14(2)3)19-16(18)15-11-7-6-8-12-15/h5-8,10-12H,1,9,13H2,2-4H3

InChI key

BTJXBZZBBNNTOV-UHFFFAOYSA-N

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C S Letizia et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 41(7), 977-981 (2003-06-14)
A toxicologic and dermatologic review of linalyl benzoate when used as a fragrance ingredient is presented.
C S Letizia et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 41(7), 989-993 (2003-06-14)
A toxicologic and dermatologic review of linalyl cinnamate when used as a fragrance ingredient is presented.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service