Skip to Content
Merck
All Photos(1)

Documents

W220604

Sigma-Aldrich

Butyl laurate

≥99%

Synonym(s):

Butyl dodecanoate, Butyl laurate, Lauric acid butyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)10COO(CH2)3CH3
CAS Number:
Molecular Weight:
256.42
FEMA Number:
2206
Beilstein:
1776360
EC Number:
Council of Europe no.:
376
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.100
NACRES:
NA.21

biological source

synthetic

reg. compliance

FDA 21 CFR 172.515

Assay

≥99%

refractive index

n20/D 1.435 (lit.)

bp

180 °C/18 mmHg (lit.)

mp

−10 °C (lit.)

density

0.855 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

waxy; soapy

SMILES string

CCCCCCCCCCCC(=O)OCCCC

InChI

1S/C16H32O2/c1-3-5-7-8-9-10-11-12-13-14-16(17)18-15-6-4-2/h3-15H2,1-2H3

InChI key

NDKYEUQMPZIGFN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neha Lamba et al.
Bioresource technology, 241, 767-774 (2017-06-21)
Biodiesel or fatty acid methyl esters (FAMEs) is primarily synthesized using edible vegetable oils and methanol with a catalyst. However, in the present study, FAMEs were synthesized from a non-edible oil (Calophyllum inophyllum also called as sura honne, Punnagam, Alexandrian

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service