All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C12H18O3
CAS Number:
Molecular Weight:
210.27
FEMA Number:
2071
EC Number:
Council of Europe no.:
2092
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.023
reg. compliance
FDA 21 CFR 172.515
Assay
≥97%
refractive index
n20/D 1.459 (lit.)
bp
258 °C (lit.)
density
0.99 g/mL at 25 °C (lit.)
SMILES string
CC(C)CCOC(=O)CCc1ccco1
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service