T70394
2,4,5-Trimethoxycinnamic acid, predominantly trans
98%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(CH3O)3C6H2CH=CHCO2H
CAS Number:
Molecular Weight:
238.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
Assay
98%
mp
164-166 °C (lit.)
SMILES string
COc1cc(OC)c(\C=C\C(O)=O)cc1OC
InChI
1S/C12H14O5/c1-15-9-7-11(17-3)10(16-2)6-8(9)4-5-12(13)14/h4-7H,1-3H3,(H,13,14)/b5-4+
InChI key
XCEGAEUDHJEYRY-SNAWJCMRSA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service