Documents
SYNV200100G
Synthware micro Dewar vacuum trap
100 mL, stopcock: glass
Synonym(s):
Synthware V200100G
About This Item
UNSPSC Code:
40161501
NACRES:
NB.51
packaging
pkg of 1 ea
manufacturer/tradename
Synthware V200100G
capacity
100 mL
Related Categories
General description
Small scale dry ice, Dewar style trap. The dry-ice chamber is 40 mm I.D.× 60 mm in length. Choice PTFE stopcock for grease-free operation.
Legal Information
Synthware is a trademark of Kemtech America Inc.