Key Documents
SYNM522405
Synthware vacuum manifold with high vacuum valves
number of ports, 5, Hose Connection: left
Synonym(s):
Synthware M522405, vacuum manifold, gas manifold
Select a Size
1 EA
€660.00
€660.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€660.00
About This Item
UNSPSC Code:
40161501
NACRES:
NB.51
€660.00
Please contact Customer Service for Availability
feature
number of ports 5
packaging
pkg of 1 ea
manufacturer/tradename
Synthware M522405
General description
With a serrated hose connection(10 mm O.D.) at left hand side. Main tube is 22 mm O.D. and the distance between valves is 100 mm center to center.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)