Documents
SYNF621410G
Synthware pressure equalizing addition funnel
10 mL, joint: ST/NS 14/20, stopcock: glass
Synonym(s):
Synthware F621410G, dropping funnel
About This Item
UNSPSC Code:
41121809
NACRES:
NB.51
Joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F621410G
capacity
10 mL
10 mL
Related Categories
General description
With glass stopcock, graduated.
Legal Information
Synthware is a trademark of Kemtech America Inc.