Skip to Content
Merck
All Photos(2)

Key Documents

P22273

Sigma-Aldrich

2-Phenylcyclohexanone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H9(=O)
CAS Number:
Molecular Weight:
174.24
Beilstein:
1368883
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

crystals

mp

56-59 °C (lit.)

SMILES string

O=C1CCCCC1c2ccccc2

InChI

1S/C12H14O/c13-12-9-5-4-8-11(12)10-6-2-1-3-7-10/h1-3,6-7,11H,4-5,8-9H2

InChI key

DRLVMOAWNVOSPE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paola Peluso et al.
Journal of chromatography. A, 1616, 460788-460788 (2019-12-24)
The halogen bond (XB) is a noncovalent interaction involving a halogen acting as electrophile and a Lewis base. In the last decades XB has found practical application in several fields. Nevertheless, despite the pivotal role of noncovalent interactions in separation
Alejandro Gran-Scheuch et al.
Frontiers in microbiology, 9, 1609-1609 (2018-08-04)
Actinobacteria are an important source of commercial (bio)compounds for the biotechnological and pharmaceutical industry. They have also been successfully exploited in the search of novel biocatalysts. We set out to explore a recently identified actinomycete, Streptomyces leeuwenhoekii C34, isolated from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service