All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C16H12O3
CAS Number:
Molecular Weight:
252.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
SMILES string
COc1ccccc1C2=CC(=O)c3ccccc3O2
InChI
1S/C16H12O3/c1-18-14-8-4-3-7-12(14)16-10-13(17)11-6-2-5-9-15(11)19-16/h2-10H,1H3
InChI key
YEHDMSUNJUONMW-UHFFFAOYSA-N
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Benjawan Wudtiwai et al.
Journal of hematology & oncology, 4, 52-52 (2011-12-22)
Tumor necrosis factor-related apoptosis-inducing ligand (TRAIL) induces apoptosis in various tumor cells, but does not affect normal cells or human leukemic cells, such as MOLT-4 and U937 cells, which are relatively resistant to TRAIL. Three flavonoids extracted from the rhizome
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service