Skip to Content
Merck
All Photos(1)

Key Documents

JKA7006

Sigma-Aldrich

4arm-PEG10K-Succinimidyl Succinate

pentaerythritol core, average Mn 10,000

Synonym(s):

4arm-PEG-Succinimidyl Succinate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C(CH2O(CH2CH2O)nCOCH2CH2COOC4H4O2N)4
UNSPSC Code:
12162002
NACRES:
NA.23

form

solid

mol wt

average Mn 10,000

reaction suitability

reactivity: amine reactive

polymer architecture

shape: 4-arm
functionality: homofunctional

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

4arm PEG Succinimidyl Succinate. Hydrogel PEG. Cleavable PEG linker. The ester linker between PEG and NHS ester enables the feature of “degradable" hydrogels.

Application

Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization

Legal Information

Product of JenKem Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Buket Ucar et al.
Acta biomaterialia, 121, 250-262 (2020-11-27)
Protection or repair of the nigrostriatal pathway represents a principal disease-modifying therapeutic strategy for Parkinson's disease (PD). Glial cell line-derived neurotrophic factor (GDNF) holds great therapeutic potential for PD, but its efficacious delivery remains difficult. The aim of this study
Buket Ucar et al.
Experimental brain research, 238(11), 2521-2529 (2020-08-31)
Vessel damage is a general pathological process in many neurodegenerative disorders, as well as spinal cord injury, stroke, or trauma. Biomaterials can present novel tools to repair and regenerate damaged vessels. The aim of the present study is to test
Kurt Moelgg et al.
Biomolecules, 11(3) (2021-04-04)
The extracellular deposition of β-amyloid (Aβ) is one of the major characteristics in Alzheimer´s disease (AD). The "spreading hypothesis" suggests that a pathological protein (similar to prions) spreads over the entire brain. The aim of the present study was to

Articles

Immune system defends against disease by hydrolyzing foreign molecules; proteases, nucleases, liver, and kidneys are involved.

Immune system defends against disease by hydrolyzing foreign molecules; proteases, nucleases, liver, and kidneys are involved.

Immune system defends against disease by hydrolyzing foreign molecules; proteases, nucleases, liver, and kidneys are involved.

Immune system defends against disease by hydrolyzing foreign molecules; proteases, nucleases, liver, and kidneys are involved.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service