Skip to Content
Merck
All Photos(2)

Key Documents

GF61123496

Silver

foil, 25mm disks, thickness 0.005mm, 99.97%

Synonym(s):

Silver, AG000130

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ag
CAS Number:
Molecular Weight:
107.87
MDL number:
UNSPSC Code:
12141740
PubChem Substance ID:
NACRES:
NA.23

Assay

99.97%

form

foil

manufacturer/tradename

Goodfellow 611-234-96

resistivity

1.59 μΩ-cm, 20°C

diam. × thickness

25 mm × 0.005 mm

bp

2212 °C (lit.)

mp

960 °C (lit.)

density

10.49 g/cm3 (lit.)

SMILES string

[Ag]

InChI

1S/Ag

InChI key

BQCADISMDOOEFD-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhiping Zhuang et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 67(2), 509-516 (2006-09-22)
The molecular geometry and vibrational frequencies of 4,4'-bipyridine (BPE) in the ground state were calculated using density functional theory (DFT) methods (B3LYP) with 6-31++G(d,p) basis set. The optimized geometric bond lengths and bond angles are obtained by DFT employing the
Ken-ichiro Matsumoto et al.
Biological & pharmaceutical bulletin, 28(11), 2029-2034 (2005-11-08)
A novel screening experiment, to find radioactive probes for non-invasive measurements of physiological functions in experimental animals, was tested using the in vivo multitracer analysis technique. The details of the efficiency of the detector settings used in the in vivo
A Rupérez et al.
Talanta, 44(2), 213-220 (1997-02-01)
Surface-enhanced Raman scattering (SERS) spectrometry of the diuretic drug triamterene is discussed. The SERS-active substrate used is a silver foil etched with nitric acid. The influence of solvent and sample doping method on sensivity, intercept and shape of the calibration
IX. Note on Silver Foil in Surgery.
J S Lewis
Annals of surgery, 50(4), 793-796 (1909-10-01)
Clinical evaluation of C-Film, a vaginal contraceptive.
O Frankman et al.
The Journal of international medical research, 3(4), 292-296 (1975-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service