GF39404561
Invar® - Controlled Expansion Alloy, Fe64Ni36
foil, 1m coil, 0.125mm thickness, half hard
Synonym(s):
FE020265
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
form
foil (coil - half hard)
manufacturer/tradename
Goodfellow 394-045-61
size × thickness
1 m × 0.125 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Legal Information
Invar is a registered trademark of Aperam Alloys Imphy
Invar is a registered trademark of Aperam Alloys Imphy
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service