Skip to Content
Merck
All Photos(1)

Documents

GF18528419

Brass alloy, Cu63Zn37

100mm tube, 3.0mm outside diameter, 0.5mm wall thickness, 2mm inside diameter, as drawn

Synonym(s):

CU027500

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
30102010
NACRES:
NA.23

form

tubes

manufacturer/tradename

Goodfellow 185-284-19

L × thickness

100 mm × 0.5 mm

O.D. × I.D.

3.0 m × 2.0 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service