Key Documents
DWKKC92812G-250
KIMBLE® KIMAX® Heavy duty volumetric wide-mouth flask with KIMCOTE® safety coating and glass stopper
glass flask, flask capacity (250 mL), class A
Select a Size
1 EACH
€821.00
€821.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EACH
€821.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
€821.00
Please contact Customer Service for Availability
material
glass flask
feature
class A
packaging
case of 6 ea
manufacturer/tradename
Kimble® KC92812G-250
flask capacity
(250 mL)
General description
KimCote Class A Heavy Duty Wide Mouth Volumetric Flasks
  • Pipet access is easy with wide-mouth volumetric flasks
  • Superior strength, durability and safety from heavy, uniform walls
  • Quick identification with large, permanent, easy-to-read markings
  • Flat interior bottoms for stirring with standard stir bars
  • Graduated to Class A volumetric tolerances
  • Standard Taper glass stoppers
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences
KIMCOTE is a registered trademark of DWK LIFE SCIENCES LLC
Kimax is a registered trademark of Gerresheimer Glass, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)