Documents
DWK73500-650
KIMBLE® disposable rimless culture tube
round bottom borosilicate glass, tube diam. (6 mm) (50 mm), non-sterile
About This Item
UNSPSC Code:
41121706
NACRES:
NB.11
material
round bottom borosilicate glass
sterility
non-sterile
packaging
case of 1000 ea shrink-wrapped trays
manufacturer/tradename
Kimble® 73500-650
tube diam.
(6 mm) (50 mm)
General description
Plain Disposable Borosilicate Glass Tubes
  • These premium quality tubes feature sturdy, uniform bottoms and consistent lengths.
  • 51 expansion borosilicate glass offers excellent chemical resistance against sodium leaching, a factor common in soda lime glass products
  • Tubes are packed in convenient trays for ease of use on bench tops and in drawers
  • Trays are shrink-wrapped in plastic film, with tubes remaining in constant alignment for protection against in-transit breakage and contamination
  • No marking spot
  • Designed from ASTM Specification E890
  • Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
Features and Benefits
• Complies with ASTM Specification E890• Manufactured from borosilicate glass 5.1• 51 expansion borosilicate glass offers excellent chemical resistance against sodium leaching, a factor common in soda lime glass products• Tubes are packed in convenient trays for ease of use on bench tops and in drawers• The 6 x 50 mm size is often referred to as a "Durham tube"• Trays are shrink-wrapped in plastic film, with tubes remaining in constant alignment for protection against in-transit breakage and contamination• No marking spot
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences