Yes, these Bottle Tags are autoclavable at 121°C.
DWK292432818
Bottle Tag, For Gl 45 Duran® Media Bottles Bottle Tag
red silicone bottle
Synonym(s):
292432818
Select a Size
€40.20
Select a Size
About This Item
€40.20
Recommended Products
material
red silicone bottle
packaging
pkg of 20 ea
manufacturer/tradename
DURAN® 292432818
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Are the silicone bottle tags autoclavable?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service