DWK231700006
Nmr Tube, 100Mhz, Borosilicate Glass 5.1, 178 Mm
Synonym(s):
231700006
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Recommended Products
material
Borosilicate glass/PTFE vial
packaging
pkg of 100 ea
manufacturer/tradename
231700006
parameter
100 MHz frequency
NMR tube size
178 mm
Application
Nuclear magnetic resonance spectroscopy (NMR spectroscopy), Storage and transport of measurement solutions Designed for use in low to most mid field NMR spectrometers, Boro 3.3, EVA
Features and Benefits
Close tolerances and accuracy, especially to their straightness, wall thickness and wall thickness distribution; Supplied with cap
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service