DWK218034403
Duran® Erlenmeyer Flask
graduated, neck Joints: (GL 32)
Synonym(s):
218034403
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Joint
Joints: neck (GL 32)
feature
graduated
packaging
pkg of 10 ea
manufacturer/tradename
DURAN® 218034403
capacity
500 mL
Looking for similar products? Visit Product Comparison Guide
Application
The flask is suitable for storage, media preparation and cultivation
Features and Benefits
Easy mixing due to conical form ; Easy-to-read scale with graduation marks to estimate the content; Fired-on, highly durable white ceramic
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service