Skip to Content
Merck
All Photos(1)

Key Documents

D68207

Sigma-Aldrich

1,2-Dichloro-3-nitrobenzene

99%

Synonym(s):

vic.-Nitro-o-dichlorobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3NO2
CAS Number:
Molecular Weight:
192.00
Beilstein:
2048029
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

99%

bp

257-258 °C (lit.)

mp

57-61 °C (lit.)

density

1.449 g/mL at 25 °C (lit.)

SMILES string

[O-][N+](=O)c1cccc(Cl)c1Cl

InChI

1S/C6H3Cl2NO2/c7-4-2-1-3-5(6(4)8)9(10)11/h1-3H

InChI key

CMVQZRLQEOAYSW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

305.6 °F

Flash Point(C)

152 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service