Skip to Content
Merck
All Photos(1)

Documents

D186406

Sigma-Aldrich

2,4-Dimethylsulfolane

95%

Synonym(s):

2,4-Dimethyltetrahydrothiophene-1,1-dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O2S
CAS Number:
Molecular Weight:
148.22
Beilstein:
111239
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

95%

refractive index

n20/D 1.473 (lit.)

bp

281 °C (lit.)

mp

−3 °C (lit.)

density

1.136 g/mL at 25 °C (lit.)

SMILES string

CC1CC(C)S(=O)(=O)C1

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

289.4 °F

Flash Point(C)

143 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service