Skip to Content
Merck
All Photos(1)

Documents

CLS8220100

Pyrex® pear-shaped graduated centrifuge tube, Goetz

capacity 100.0 mL, O.D. × L 58 mm × 159 mm

Synonym(s):

Corning® 8220

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121703
NACRES:
NB.43

material

borosilicate glass
round bottom tube

sterility

non-sterile

packaging

case of 12
pack of 12

manufacturer/tradename

Corning 8220-100

O.D. × L

58 mm × 159 mm

capacity

100.0 mL

color

clear

suitability

suitable for (phosphorus determinations in iron and steel)

General description

The lower stem is graduated upward to 1 mL in 0.05 mL divisions. The body is graduated from 1 mL to 5 mL in 1 mL divisions; from 5 mL to 50 mL in 5 mL divisions; and from 50 mL to 100 mL in 10 mL divisions.

Legal Information

Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service