CLS6180100
Pyrex® fluted funnel, short stem
stem L 100 mm
Synonym(s):
Corning® 6180
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
manufacturer/tradename
Corning 6180-100
stem L
100 mm
stem O.D.
8-9 mm
top I.D.
100 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Beaded edge. Has depressed inside flutings to decrease filtering time. 60° angle cone.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service