Skip to Content
Merck
All Photos(1)

Key Documents

C97806

Sigma-Aldrich

Cyclododecylamine

97%

Synonym(s):

Aminocyclododecane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C12H23NH2
CAS Number:
Molecular Weight:
183.33
Beilstein:
2072978
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

bp

122-124 °C/7 mmHg (lit.)

mp

27-29 °C (lit.)

SMILES string

NC1CCCCCCCCCCC1

InChI

1S/C12H25N/c13-12-10-8-6-4-2-1-3-5-7-9-11-12/h12H,1-11,13H2

InChI key

HBGGBCVEFUPUNY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karthik Nadendla et al.
Molecular pharmaceutics, 16(7), 2922-2928 (2019-05-24)
We have previously described the photoactivated depot (PAD) approach for the light-stimulated release of therapeutic proteins such as insulin. The aim of this method is to release insulin from a shallow dermal depot in response to blood glucose information, using
Hilary M Corkran et al.
Bioorganic & medicinal chemistry, 24(4), 693-702 (2016-01-27)
A series of glycitylamines, which were prepared in few steps from readily available carbohydrates, were tested for their ability to inhibit tuberculosis growth in an Alamar Blue BCG colourimetric assay. Several derivatives, including (2R,3R)-1-(hexadecylamine)pent-4-ene-2,3-diol, (2R,3R)-1-(hexadecylmethylamino)pent-4-ene-2,3-diol and 5-deoxy-5-hexadecylmethylamino-D-arabinitol, were prepared in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service