C5009
Ethyl 2-methyl-4-oxo-2-cyclohexenecarboxylate
90%, technical grade
Synonym(s):
4-Carbethoxy-3-methyl-2-cyclohexen-1-one, Hagemann’s ester
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C10H14O3
CAS Number:
Molecular Weight:
182.22
Beilstein:
1371741
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
grade
technical grade
Assay
90%
form
liquid
refractive index
n20/D 1.488 (lit.)
bp
268-272 °C (lit.)
density
1.078 g/mL at 25 °C (lit.)
SMILES string
CCOC(=O)C1CCC(=O)C=C1C
InChI
1S/C10H14O3/c1-3-13-10(12)9-5-4-8(11)6-7(9)2/h6,9H,3-5H2,1-2H3
InChI key
VLTANIMRIRCCOQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service