Skip to Content
Merck
All Photos(1)

Key Documents

BMSW3120

Benchmark Compact Anti Static Ionizer

AC/DC input 115 V

Synonym(s):

ionizer, anti static, balance, accuris

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€1,420.00

€1,420.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EACH
€1,420.00

About This Item

UNSPSC Code:
41103400
NACRES:
NA.74

€1,420.00


Please contact Customer Service for Availability

manufacturer/tradename

Benchmark Scientific W3120

Looking for similar products? Visit Product Comparison Guide

General description

Compact Anti Static Ionizer

Application

Removing static from balances

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service