Skip to Content
Merck
All Photos(1)

Documents

BMSE1101SKEUK

Benchmark myGel Mini Electrophoresis System Starter Kit

AC/DC input 230 V, UK plug

Synonym(s):

protein separation, gel catsing

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105300
NACRES:
NB.13

manufacturer/tradename

Benchmark Scientific E1101-SK-E-UK

Looking for similar products? Visit Product Comparison Guide

General description

myGel Mini Electrophoresis Starter kit includes mini gel box with integrated power supply, casting set, small bottle of agarose and a mini balance

Application

Electrophoresis

Legal Information

myGel is a trademark of Benchmark Scientific, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service