BAF378720900
SP Bel-Art® Sterileware™ Jumbo Sampling Pipettes
capacity × L 180 mL (6⅛ oz) × 35.5 in. (90 cm), pack of 50 ea (individually wrapped)
Synonym(s):
pipettes
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
Recommended Products
material
high-density polyethylene
packaging
pack of 50 ea (individually wrapped)
manufacturer/tradename
Bel-Art F37872-0900
O.D.
0.875 in. (21 mm)
21 mm
bore size
0.125 in. (3 mm)
capacity × L
180 mL (6⅛ oz) × 35.5 in. (90 cm)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
These sterile Jumbo Pipettes are ideal for sampling from narrow- or wide-mouth containers up to 208-liter (55 Gallon) drums.
Features and Benefits
These sterile spoons are ideal for sampling and dispensing of powders, granules and liquids.
Note: Sterilized products are not returnable.
These sampling spoons are designed for retrieving samples from a larger source. Capacity/volumes are estimated and are not meant for taking exact sample weights/volumes. Stated volumes represent typical heaping samples.
- Long handle allows sampling from a distance reducing the risk of contamination
- Handle is bent at bowl to facilitate easier retrieval of samples, including from within tall containers
- Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
- High density polyethylene (HDPE) spoons heat sealed in individual polyethylene bags
- Gamma irradiated sterile (SAL 10^-6)
- Lot stamped for accurate tracing
Note: Sterilized products are not returnable.
These sampling spoons are designed for retrieving samples from a larger source. Capacity/volumes are estimated and are not meant for taking exact sample weights/volumes. Stated volumes represent typical heaping samples.
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Sterileware is a trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service